COMH4211775D Combi-Blocks, Inc.

3-Azetidin-3-yl-1-methyl-1h-pyrazolo[3,4-b]pyridine trifluoroacetate

  • MDL number MFCD31537589
  • Cas Number : 2201364-76-1
  • Molecular Weight : 302.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8198

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8198
Availability details for
Quantity
  • Availability
  • Add to Cart