COMH4211775F Combi-Blocks, Inc.

Bis-sulfone-peg4-methyltetrazine

  • MDL number MFCD29052190
  • Molecular Weight : 883.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8216
Availability details for
Quantity
  • Availability
  • Add to Cart