COMH42117763 Combi-Blocks, Inc.

1-(2-Fluorobenzyl)-1h-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD08445753
  • Cas Number : 106308-42-3
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8229

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8229
Availability details for
Quantity
  • Availability
  • Add to Cart