COMH42117764 Combi-Blocks, Inc.

5-Allyl-2,6-dimethyl-4-pyrimidinol

  • MDL number MFCD00461655
  • Cas Number : 78304-54-8
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8230

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8230
Availability details for
Quantity
  • Availability
  • Add to Cart