COMH4211776C Combi-Blocks, Inc.

(2E)-3-(Dimethylamino)-1-(3,4-dimethylphenyl)prop-2-en-1-one

  • MDL number MFCD14449819
  • Cas Number : 72851-83-3
  • Molecular Weight : 203.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8274
Availability details for
Quantity
  • Availability
  • Add to Cart