COMH42117771 Combi-Blocks, Inc.

2-[2-(1,3-Dioxo-1,3-dihydro-2h-isoindol-2-yl)ethoxy]-4,6-dimethylnicotinonitrile

  • MDL number MFCD31583434
  • Cas Number : 2270913-28-3
  • Molecular Weight : 321.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8292
Availability details for
Quantity
  • Availability
  • Add to Cart