COMH42117773 Combi-Blocks, Inc.

4-[(3,5-Dimethylisoxazol-4-yl)methyl]-5-methylisoxazole-3-carboxylic acid hydrate

  • MDL number MFCD27756337
  • Molecular Weight : 254.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8311
Availability details for
Quantity
  • Availability
  • Add to Cart