COMH42117777 Combi-Blocks, Inc.

(2-Fluoro-4-methoxyphenyl)(5-hydroxy-1-benzofuran-3-yl)methanone

  • MDL number MFCD30188505
  • Cas Number : 2108836-36-6
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8341
Availability details for
Quantity
  • Availability
  • Add to Cart