COMH42117778 Combi-Blocks, Inc.

2-Amino-n,n-diethyl-3-methylbenzamide

  • MDL number MFCD09043883
  • Cas Number : 926250-76-2
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8347
Availability details for
Quantity
  • Availability
  • Add to Cart