COMH42117787 Combi-Blocks, Inc.

5-Methyl-n-phenyl-4,7-dihydro[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxamide

  • MDL number MFCD26122012
  • Cas Number : 1432144-28-9
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8401

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8401
Availability details for
Quantity
  • Availability
  • Add to Cart