COMH42117792 Combi-Blocks, Inc.

3-Nitro-4-(1-pyrrolidinyl)-pyridine

  • MDL number MFCD30293503
  • Cas Number : 90674-35-4
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8464
Availability details for
Quantity
  • Availability
  • Add to Cart