COMH42117795 Combi-Blocks, Inc.

C-(1-Phenyl-cyclopentyl)-methylamine hydrochloride

  • MDL number MFCD01211603
  • Cas Number : 17380-61-9
  • Molecular Weight : 211.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8483
Availability details for
Quantity
  • Availability
  • Add to Cart