COMH42117796 Combi-Blocks, Inc.

[(8-Methyl-3-oxo-3,4-dihydro-2h-1,4-benzoxazin-7-yl)oxy]acetic acid

  • MDL number MFCD31583452
  • Cas Number : 2270906-74-4
  • Molecular Weight : 237.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-((8-methyl-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl)oxy)acetic acid
Inchi Code 1S/C11H11NO5/c1-6-8(16-5-10(14)15)3-2-7-11(6)17-4-9(13)12-7/h2-3H,4-5H2,1H3,(H,12,13)(H,14,15)
Inchi Key NGOZWWANFNWJQE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8492

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8492
Availability details for
Quantity
  • Availability
  • Add to Cart