COMH42117797 Combi-Blocks, Inc.

2-Amino-5-(3-fluorobenzyl)-6-methylpyrimidin-4(3h)-one

  • MDL number MFCD31581343
  • Cas Number : 2270913-31-8
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8497
Availability details for
Quantity
  • Availability
  • Add to Cart