COMH42117798 Combi-Blocks, Inc.

tert-Butyl 4-hydroxytetrahydrofuran-3-carboxylate

  • MDL number MFCD28363986
  • Cas Number : 797038-33-6
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8501

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8501
Availability details for
Quantity
  • Availability
  • Add to Cart