COMH4211779D Combi-Blocks, Inc.

5-Methoxy-2-piperidin-4-yl-1h-benzimidazole dihydrochloride

  • MDL number MFCD31583424
  • Cas Number : 2322533-24-2
  • Molecular Weight : 304.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C13H17N3O.2ClH/c1-17-10-2-3-11-12(8-10)16-13(15-11)9-4-6-14-7-5-9;;/h2-3,8-9,14H,4-7H2,1H3,(H,15,16);2*1H
Inchi Key QZFJMPDDONGGTR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8519

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8519
Availability details for
Quantity
  • Availability
  • Add to Cart