COMH421177A2 Combi-Blocks, Inc.

(E)-6-(4,6-Dimethoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

  • MDL number MFCD22375170
  • Cas Number : 38877-93-9
  • Molecular Weight : 334.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8560
Availability details for
Quantity
  • Availability
  • Add to Cart