COMH421177A3 Combi-Blocks, Inc.

1-(2-Amino-7-methyl[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)ethanone

  • MDL number MFCD31581335
  • Cas Number : 2270905-51-4
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8564
Availability details for
Quantity
  • Availability
  • Add to Cart