COMH421177A4 Combi-Blocks, Inc.

4-Ethoxy-n-methyl-2-nitroaniline

  • MDL number MFCD11855379
  • Cas Number : 61679-18-3
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-ethoxy-N-methyl-2-nitroaniline
Inchi Code 1S/C9H12N2O3/c1-3-14-7-4-5-8(10-2)9(6-7)11(12)13/h4-6,10H,3H2,1-2H3
Inchi Key DTJXSLYJMZSTII-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8565

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8565
Availability details for
Quantity
  • Availability
  • Add to Cart