COMH421177A6 Combi-Blocks, Inc.

M-Peg8-(ch2)12-phosphonic acid

  • MDL number MFCD30723266
  • Cas Number : 2093153-86-5
  • Molecular Weight : 588.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8570

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8570
Availability details for
Quantity
  • Availability
  • Add to Cart