COMH421177A7 Combi-Blocks, Inc.

[(2-Formyl-1-methyl-1h-indol-3-yl)thio]acetic acid

  • MDL number MFCD14281961
  • Cas Number : 885123-03-5
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8579
Availability details for
Quantity
  • Availability
  • Add to Cart