COMH421177B5 Combi-Blocks, Inc.

Methyl 5-[(e)-2-(dimethylamino)vinyl][1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD31619050
  • Cas Number : 2279136-03-5
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8658
Availability details for
Quantity
  • Availability
  • Add to Cart