COMH421177B8 Combi-Blocks, Inc.

17-(5-Amino-1h-1,2,4-triazol-3-yl)heptadecan-7-ol

  • MDL number MFCD31619047
  • Cas Number : 2279123-46-3
  • Molecular Weight : 338.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8665

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8665
Availability details for
Quantity
  • Availability
  • Add to Cart