COMH421177BB Combi-Blocks, Inc.

T-Boc-aminooxy-peg3-thiol

  • MDL number MFCD29764338
  • Cas Number : 1895922-75-4
  • Molecular Weight : 325.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8674
Availability details for
Quantity
  • Availability
  • Add to Cart