COMH421177BF Combi-Blocks, Inc.

6,6-Dimethyloxane-2-carboxylic acid

  • MDL number MFCD29954553
  • Cas Number : 1849376-14-2
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8695

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8695
Availability details for
Quantity
  • Availability
  • Add to Cart