COMH421177CA Combi-Blocks, Inc.

Bis-sulfone-peg12-nhs ester

  • MDL number MFCD30458026
  • Cas Number : 2055105-51-4
  • Molecular Weight : 1197.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C56H80N2O22S2/c1-45-3-11-50(12-4-45)81(64,65)43-49(44-82(66,67)51-13-5-46(2)6-14-51)55(62)47-7-9-48(10-8-47)56(63)57-18-20-69-22-24-71-26-28-73-30-32-75-34-36-77-38-40-79-42-41-78-39-37-76-35-33-74-31-29-72-27-25-70-23-21-68-19-17-54(61)80-58-52(59)15-16-53(58)60/h3-14,49H,15-44H2,1-2H3,(H,57,63)
Inchi Key BJDSAKOWMGLMII-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8735
Availability details for
Quantity
  • Availability
  • Add to Cart