COMH421177D3 Combi-Blocks, Inc.

Tfp-peg13-dspe

  • MDL number MFCD28118975
  • Molecular Weight : 1568.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8775
Availability details for
Quantity
  • Availability
  • Add to Cart