COMH421177D6 Combi-Blocks, Inc.

3-Aminooxolane-3-carbonitrile

  • MDL number MFCD18333627
  • Cas Number : 1037301-15-7
  • Molecular Weight : 112.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8784

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8784
Availability details for
Quantity
  • Availability
  • Add to Cart