COMH421177D8 Combi-Blocks, Inc.

Diethyl-(2-(2-[2-(2-mercapto-ethoxy)-ethoxy]-ethoxy)-ethyl)phosphonate

  • MDL number MFCD21596688
  • Cas Number : 1360716-43-3
  • Molecular Weight : 330.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8786
Availability details for
Quantity
  • Availability
  • Add to Cart