COMH421177D9 Combi-Blocks, Inc.

6-Isopropyl-1,3-dimethyl-1h-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09040584
  • Cas Number : 929973-66-0
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8790
Availability details for
Quantity
  • Availability
  • Add to Cart