COMH421177DE Combi-Blocks, Inc.

3-Cyclopropyl-1-methyl-1h-pyrazole-5-carbohydrazide

  • MDL number MFCD30901683
  • Cas Number : 2168958-23-2
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8818
Availability details for
Quantity
  • Availability
  • Add to Cart