COMH421177E9 Combi-Blocks, Inc.

2H-3,4'-Bi-1,2,4-triazole

  • MDL number MFCD03306875
  • Cas Number : 68984-29-2
  • Molecular Weight : 136.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8876

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8876
Availability details for
Quantity
  • Availability
  • Add to Cart