COMH421177EC Combi-Blocks, Inc.

2-[(1E)-(2-Chloro-5-nitrophenyl)methylene]aminoethanol

  • MDL number MFCD31583436
  • Cas Number : 2270915-09-6
  • Molecular Weight : 228.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8884

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8884
Availability details for
Quantity
  • Availability
  • Add to Cart