COMH42119F0E Combi-Blocks, Inc.

3H-1,2,3-triazole-4-carboxylic acid hydrochloride

  • MDL number MFCD32065684
  • Cas Number : 2365418-30-8
  • Molecular Weight : 149.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4544
Availability details for
Quantity
  • Availability
  • Add to Cart