COMH42119F11 Combi-Blocks, Inc.

Methyl 4-[N-(2-ethoxy-2-oxoethyl)methanesulfonamido]-3-nitrobenzoate

  • MDL number MFCD32065700
  • Cas Number : 2365418-81-9
  • Molecular Weight : 360.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2494
Availability details for
Quantity
  • Availability
  • Add to Cart