COMH42119F12 Combi-Blocks, Inc.

5-Iodo-2,4-dimethoxyquinazoline

  • MDL number MFCD32065703
  • Cas Number : 2365419-26-5
  • Molecular Weight : 316.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2530
Availability details for
Quantity
  • Availability
  • Add to Cart