COMH42119F14 Combi-Blocks, Inc.

6-bromo-N-isopropyl-3-nitropyridin-2-amine

  • MDL number MFCD32065705
  • Cas Number : 2365418-56-8
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2534

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2534
Availability details for
Quantity
  • Availability
  • Add to Cart