COMH42119F15 Combi-Blocks, Inc.

6-Bromo-2-cyclopropylamino-3-nitropyridine

  • MDL number MFCD32065706
  • Cas Number : 2365418-31-9
  • Molecular Weight : 258.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-N-cyclopropyl-3-nitropyridin-2-amine
Inchi Code 1S/C8H8BrN3O2/c9-7-4-3-6(12(13)14)8(11-7)10-5-1-2-5/h3-5H,1-2H2,(H,10,11)
Inchi Key ZAOABXZZICCZTI-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2535
Availability details for
Quantity
  • Availability
  • Add to Cart