COMH42119F17 Combi-Blocks, Inc.

tert-Butyl 4-(2-chloro-3-ethoxy-3-oxopropanoyl)piperidine-1-carboxylate

  • MDL number MFCD32065709
  • Cas Number : 2365419-13-0
  • Molecular Weight : 333.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4568

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4568
Availability details for
Quantity
  • Availability
  • Add to Cart