COMH42119F18 Combi-Blocks, Inc.

Ethyl 4-bromo-7-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-2-carboxylate

  • MDL number MFCD32065718
  • Cas Number : 2377609-01-1
  • Molecular Weight : 394.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4718
Availability details for
Quantity
  • Availability
  • Add to Cart