COMH42119F19 Combi-Blocks, Inc.

8-(oxolan-2-ylmethoxy)quinolin-5-ol

  • MDL number MFCD32065720
  • Cas Number : 2365419-68-5
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4724
Availability details for
Quantity
  • Availability
  • Add to Cart