COMH42119F1B Combi-Blocks, Inc.

6-Chloro-5-(methylamino)pyridin-3-ol

  • MDL number MFCD32065727
  • Cas Number : 2365418-83-1
  • Molecular Weight : 158.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4755
Availability details for
Quantity
  • Availability
  • Add to Cart