COMH42119F1C Combi-Blocks, Inc.

4-benzyloxycyclohexyl bromide

  • MDL number MFCD32065730
  • Cas Number : 2365418-33-1
  • Molecular Weight : 269.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4759
Availability details for
Quantity
  • Availability
  • Add to Cart