COMH42119F1D Combi-Blocks, Inc.

7-Bromo-8-methoxyquinazolin-2-amine

  • MDL number MFCD32065731
  • Cas Number : 2365420-01-3
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4764
Availability details for
Quantity
  • Availability
  • Add to Cart