COMH42119F1E Combi-Blocks, Inc.

1-(2-Bromo-4-chloro-6-methylphenyl)piperazine

  • MDL number MFCD32065732
  • Cas Number : 2365418-97-7
  • Molecular Weight : 289.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C11H14BrClN2/c1-8-6-9(13)7-10(12)11(8)15-4-2-14-3-5-15/h6-7,14H,2-5H2,1H3
Inchi Key RCOBYXAAJLMLHB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4766
Availability details for
Quantity
  • Availability
  • Add to Cart