COMH42119F22 Combi-Blocks, Inc.

tert-Butyl N-(2-bromo-5-methoxyphenyl)-N-[(tert-butoxy)carbonyl]carbamate

  • MDL number MFCD32065738
  • Cas Number : 2365418-98-8
  • Molecular Weight : 402.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4777
Availability details for
Quantity
  • Availability
  • Add to Cart