COMH42119F23 Combi-Blocks, Inc.

1,3-Dimethyl 2-(4-methoxypyrimidin-2-yl)propanedioate

  • MDL number MFCD32065739
  • Cas Number : 2365418-38-6
  • Molecular Weight : 240.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4783

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4783
Availability details for
Quantity
  • Availability
  • Add to Cart