COMH42119F24 Combi-Blocks, Inc.

Methyl 5-bromo-2-{[N-(1-methoxy-1-oxopropan-2-yl)methanesulfonamido]methyl}benzoate

  • MDL number MFCD32065740
  • Molecular Weight : 408.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-2-((N-(1-methoxy-1-oxopropan-2-yl)methylsulfonamido)methyl)benzoate
Inchi Code 1S/C14H18BrNO6S/c1-9(13(17)21-2)16(23(4,19)20)8-10-5-6-11(15)7-12(10)14(18)22-3/h5-7,9H,8H2,1-4H3
Inchi Key JFAXRFAMVYEIDK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4784

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4784
Availability details for
Quantity
  • Availability
  • Add to Cart