COMH42119F26 Combi-Blocks, Inc.

2,2-Dimethylpropyl 5-bromopyridine-3-sulfonate

  • MDL number MFCD32065742
  • Cas Number : 2365419-14-1
  • Molecular Weight : 308.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4790
Availability details for
Quantity
  • Availability
  • Add to Cart