COMH42119F27 Combi-Blocks, Inc.

(2E)-3-(2-Fluorophenyl)-2-(2-hydroxyphenyl)prop-2-enoic acid

  • MDL number MFCD32065743
  • Cas Number : 2365421-26-5
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4792
Availability details for
Quantity
  • Availability
  • Add to Cart